Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108865.1 | NP_001102335.1 | 1 - 2 | 277 | 75 | |
GenBank mRNA | AB002591.1 | - | 1 - 2 | 528 | 75 | |
FQ215794.1 | - | 1 - 2 | 271 | 75 | ||
FQ229094.1 | - | 1 - 2 | 259 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |