Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147210.2 | NP_671743.2 | 3 - 4 | 576 | 156 | |
GenBank mRNA | U15660.1 | - | 2 - 3 | 293 | 156 | |
U20796.1 | - | 2 - 3 | 233 | 156 | ||
X78135.1 | - | 3 - 4 | 429 | 156 | ||
X82777.1 | - | 3 - 4 | 511 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Rattus norvegicus Probe spans exons |