Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271381.1 | NP_001258310.1 | 7 - 8 | 561 | 78 | |
XM_008772413.1 | XP_008770635.1 | 6 - 7 | 726 | 78 | ||
GenBank mRNA | BC168970.1 | - | 6 - 7 | 923 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |