Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022634.2 | NP_072156.2 | 2 - 3 | 89 | 110 | |
XM_006256078.2 | XP_006256140.1 | 2 - 3 | 230 | 110 | ||
XM_006256079.2 | XP_006256141.1 | 2 - 3 | 230 | 110 | ||
GenBank mRNA | AF208230.1 | - | 2 - 3 | 89 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |