Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032079.2 | NP_114468.2 | 7 - 8 | 937 | 107 | |
GenBank mRNA | BC087010.1 | - | 7 - 8 | 937 | 7191358 | 107 |
U95727.1 | - | 7 - 8 | 971 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |