Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170787.2 | NP_740768.1 | 14 - 15 | 2457 | 77 | |
GenBank mRNA | AF541925.1 | - | 14 - 15 | 2457 | 77 | |
AY049038.1 | - | 13 - 14 | 2623 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |