Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172066.1 | NP_742063.1 | 3 - 4 | 954 | 65 | |
GenBank mRNA | BC086981.1 | - | 3 - 4 | 920 | 7112403 | 65 |
Y16774.1 | - | 3 - 4 | 954 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |