Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270556.1 | NP_001257485.1 | 12 - 13 | 1683 | 111 | |
NM_012806.2 | NP_036938.2 | 12 - 13 | 1710 | 111 | ||
XM_008769992.1 | XP_008768214.1 | 11 - 12 | 4502 | 111 | ||
GenBank mRNA | DQ377224.1 | - | 9 - 10 | 1057 | 111 | |
FQ211777.1 | - | 12 - 13 | 1682 | 111 | ||
L27128.1 | - | 12 - 13 | 1420 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |