Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021739.2 | NP_068507.2 | 16 - 17 | 1262 | 140 | |
XM_006251177.2 | XP_006251239.1 | 15 - 16 | 1269 | 140 | ||
GenBank mRNA | M16112.1 | - | 16 - 17 | 1242 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Protein Phosphatase Rattus norvegicus Probe spans exons |