Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024155.3 | NP_077069.3 | 8 - 9 | 616 | 79 | |
XM_006236841.2 | XP_006236903.1 | 9 - 10 | 703 | 79 | ||
XM_006236842.2 | XP_006236904.1 | 8 - 9 | 1239 | 79 | ||
GenBank mRNA | BC085688.1 | - | 8 - 9 | 616 | 7191458 | 79 |
D38224.2 | - | 6 - 7 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |