Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024391.1 | NP_077367.1 | 3 - 4 | 717 | 126 | |
GenBank mRNA | BC088134.1 | - | 3 - 4 | 708 | 7326035 | 126 |
X91234.1 | - | 3 - 4 | 717 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |