Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053495.1 | NP_445947.1 | 3 - 4 | 340 | 80 | |
XM_008767042.1 | XP_008765264.1 | 5 - 6 | 864 | 80 | ||
XM_008767043.1 | XP_008765265.1 | 4 - 5 | 750 | 80 | ||
XM_008767044.1 | XP_008765266.1 | 4 - 5 | 747 | 80 | ||
GenBank mRNA | AB002801.1 | - | 3 - 4 | 340 | 80 | |
AJ272428.1 | - | 5 - 6 | 627 | 80 | ||
AJ272429.1 | - | 4 - 5 | 513 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |