Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173109.1 | NP_775132.1 | 1 - 2 | 119 | 64 | |
XM_008762473.1 | XP_008760695.1 | 1 - 2 | 124 | 64 | ||
GenBank mRNA | AJ515239.1 | - | 1 - 2 | 119 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |