Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270871.1 | NP_001257800.1 | 8 - 9 | 456 | 97 | |
NM_001270872.1 | NP_001257801.1 | 7 - 8 | 420 | 97 | ||
NM_181366.2 | NP_852031.1 | 9 - 10 | 504 | 97 | ||
XM_006256916.2 | XP_006256978.1 | 9 - 10 | 544 | 97 | ||
XM_008773154.1 | XP_008771376.1 | 8 - 9 | 884 | 97 | ||
XM_008773155.1 | XP_008771377.1 | 8 - 9 | 494 | 97 | ||
XM_008773156.1 | XP_008771378.1 | 7 - 8 | 848 | 97 | ||
XM_008773157.1 | XP_008771379.1 | 7 - 8 | 842 | 97 | ||
XM_008773158.1 | XP_008771380.1 | 6 - 7 | 806 | 97 | ||
XM_008773159.1 | XP_008771381.1 | 5 - 6 | 397 | 97 | ||
XM_008773160.1 | XP_008771382.1 | 8 - 9 | 885 | 97 | ||
GenBank mRNA | AF538953.1 | - | 9 - 10 | 514 | 97 | |
AF538958.1 | - | 8 - 9 | 466 | 97 | ||
AF538959.1 | - | 7 - 8 | 430 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |