Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031081.1 | NP_112343.1 | 9 - 10 | 1014 | 168 | |
XM_006246036.2 | XP_006246098.1 | 9 - 10 | 2866 | 168 | ||
XM_006246038.2 | XP_006246100.1 | 9 - 10 | 1076 | 168 | ||
GenBank mRNA | Y15748.1 | - | 9 - 10 | 1014 | 168 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |