Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022385.3 | NP_071780.1 | 1 - 2 | 96 | 101 | |
GenBank mRNA | BC061553.1 | - | 179 | 6889396 | 101 | |
FQ226660.1 | - | 1 - 2 | 152 | 101 | ||
X76920.1 | - | 1 - 2 | 96 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |