Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080584.4 | NP_542151.1 | 6 - 7 | 987 | 90 | |
XM_008759855.1 | XP_008758077.1 | 7 - 8 | 810 | 90 | ||
XM_008759856.1 | XP_008758078.1 | 7 - 8 | 928 | 90 | ||
GenBank mRNA | BC107906.1 | - | 6 - 7 | 847 | 90 | |
BC128715.1 | - | 6 - 7 | 916 | 7456955 | 90 | |
M73808.1 | - | 6 - 7 | 987 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |