Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramel6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109342.1 | NP_001102812.1 | 3 - 4 | 1130 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |