Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173973.1 | NP_001167444.1 | 11 - 12 | 1718 | 78 | |
XM_008761518.1 | XP_008759740.1 | 10 - 11 | 1234 | 78 | ||
GenBank mRNA | BC158723.1 | - | 6 - 7 | 715 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |