Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aknad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044303.2 | NP_001037768.2 | 4 - 5 | 1274 | 66 | |
NR_073134.1 | 4 - 5 | 1274 | 66 | |||
XM_008761422.1 | XP_008759644.1 | 4 - 5 | 2455 | 66 | ||
GenBank mRNA | BC079051.1 | - | 4 - 5 | 1274 | 7115381 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |