Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077645.1 | NP_001071113.1 | 3 - 4 | 286 | 94 | |
XM_006233451.2 | XP_006233513.1 | 3 - 4 | 1003 | 94 | ||
XM_006233452.2 | XP_006233514.1 | 3 - 4 | 2508 | 94 | ||
XM_006233453.2 | XP_006233515.1 | 3 - 4 | 750 | 94 | ||
GenBank mRNA | D10770.1 | - | 3 - 4 | 286 | 94 | |
X53261.1 | - | 2 - 3 | 181 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |