Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233529.2 | XP_006233591.1 | 3 - 4 | 465 | 106 | |
XM_575065.5 | XP_575065.4 | 3 - 4 | 465 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |