Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012219.1 | NP_001012219.1 | 5 - 6 | 753 | 66 | |
XM_006233528.2 | XP_006233590.1 | 6 - 7 | 1019 | 66 | ||
GenBank mRNA | BC090011.1 | - | 5 - 6 | 753 | 7324565 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |