Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134716.1 | NP_001128188.1 | 729 | 86 | ||
GenBank mRNA | BC168186.1 | - | 729 | 7100019 | 86 | |
GU357484.1 | - | 528 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |