Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127635.2 | NP_001121107.1 | 3 - 4 | 273 | 96 | |
GenBank mRNA | BC166539.1 | - | 3 - 4 | 273 | 7191896 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |