Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107361.1 | NP_001100831.1 | 13 - 14 | 1531 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |