Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107974.1 | NP_001101444.1 | 4 - 5 | 789 | 141 | |
GenBank mRNA | FQ211274.1 | - | 530 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |