Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077671.1 | NP_001071139.1 | 4 - 5 | 910 | 73 | |
XM_006238865.2 | XP_006238927.1 | 3 - 4 | 1540 | 73 | ||
GenBank mRNA | BC127516.1 | - | 4 - 5 | 910 | 7102954 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |