Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134619.1 | NP_001128091.1 | 4 - 5 | 2829 | 59 | |
XM_006235428.2 | XP_006235490.1 | 5 - 6 | 2906 | 59 | ||
XM_008762375.1 | XP_008760597.1 | 5 - 6 | 3669 | 59 | ||
GenBank mRNA | FQ216244.1 | - | 4 - 5 | 542 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |