Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024764.1 | NP_001019935.1 | 6 - 7 | 721 | 79 | |
XM_008767685.1 | XP_008765907.1 | 7 - 8 | 701 | 79 | ||
GenBank mRNA | BC097463.1 | - | 6 - 7 | 721 | 7457579 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |