Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108297.1 | NP_001101767.1 | 6 - 7 | 800 | 111 | |
XM_006247347.1 | XP_006247409.1 | 6 - 7 | 835 | 111 | ||
GenBank mRNA | BC112385.1 | - | 5 - 6 | 1670 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |