Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013183.1 | NP_001013201.1 | 6 - 7 | 690 | 72 | |
GenBank mRNA | BC091240.1 | - | 6 - 7 | 690 | 7325087 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |