Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047842.1 | NP_001041307.1 | 12 - 13 | 1647 | 78 | |
XM_006246469.2 | XP_006246531.1 | 9 - 10 | 1430 | 78 | ||
GenBank mRNA | AY383687.1 | - | 12 - 13 | 1647 | 78 | |
BC099219.1 | - | 5 - 6 | 661 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |