Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105767.1 | NP_001099237.1 | 9 - 10 | 1335 | 106 | |
XM_008767550.1 | XP_008765772.1 | 8 - 9 | 1540 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |