Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270871.1 | NP_001257800.1 | 1 - 2 | 50 | 70 | |
NM_001270872.1 | NP_001257801.1 | 1 - 2 | 50 | 70 | ||
NM_181366.2 | NP_852031.1 | 1 - 2 | 50 | 70 | ||
GenBank mRNA | AF538953.1 | - | 1 - 2 | 60 | 70 | |
AF538958.1 | - | 1 - 2 | 60 | 70 | ||
AF538959.1 | - | 1 - 2 | 60 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |