Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039101.1 | NP_001034190.1 | 7 - 8 | 806 | 72 | |
XM_006257145.2 | XP_006257207.1 | 7 - 8 | 863 | 72 | ||
XM_008773347.1 | XP_008771569.1 | 8 - 9 | 1626 | 72 | ||
XM_008773348.1 | XP_008771570.1 | 9 - 10 | 779 | 72 | ||
GenBank mRNA | AY886531.1 | - | 7 - 8 | 806 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |