Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134568.1 | NP_001128040.1 | 180 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |