Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108499.1 | NP_001101969.1 | 3 - 4 | 373 | 81 | |
XM_006230081.2 | XP_006230143.1 | 3 - 4 | 1384 | 81 | ||
XM_006230082.2 | XP_006230144.1 | 3 - 4 | 1384 | 81 | ||
XM_006230084.2 | XP_006230146.1 | 3 - 4 | 1163 | 81 | ||
XM_008759769.1 | XP_008757991.1 | 3 - 4 | 1384 | 81 | ||
GenBank mRNA | BC168986.1 | - | 2 - 3 | 791 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |