Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_342213.6 | XP_342214.4 | 2 - 3 | 128 | 78 | |
GenBank mRNA | FQ229099.1 | - | 2 - 3 | 114 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |