Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130433.1 | NP_569117.1 | 2 - 3 | 348 | 66 | |
GenBank mRNA | FQ216726.1 | - | 2 - 3 | 211 | 66 | |
FQ228855.1 | - | 2 - 3 | 213 | 66 | ||
X05341.1 | - | 2 - 3 | 348 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |