Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105910.1 | NP_001099380.1 | 18 - 19 | 2769 | 69 | |
XM_006248865.2 | XP_006248927.1 | 18 - 19 | 2845 | 69 | ||
XM_006248866.2 | XP_006248928.1 | 19 - 20 | 3437 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |