Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107647.2 | NP_001101117.1 | 10 - 11 | 1330 | 91 | |
XM_006231836.2 | XP_006231898.1 | 10 - 11 | 1758 | 91 | ||
XM_008760671.1 | XP_008758893.1 | 9 - 10 | 3953 | 91 | ||
XM_008760672.1 | XP_008758894.1 | 10 - 11 | 3805 | 91 | ||
XM_008760673.1 | XP_008758895.1 | 10 - 11 | 1562 | 91 | ||
GenBank mRNA | BC107908.1 | - | 77 | 91 | ||
BC131851.1 | - | 10 - 11 | 1330 | 7189311 | 91 | |
FQ211666.1 | - | 10 - 11 | 1381 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |