Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053494.2 | NP_445946.2 | 5 - 6 | 754 | 73 | |
XM_008761779.1 | XP_008760001.1 | 5 - 6 | 795 | 73 | ||
GenBank mRNA | AJ245935.1 | - | 5 - 6 | 754 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |