Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106562.1 | NP_001100032.1 | 3 - 4 | 544 | 90 | |
XM_006233757.2 | XP_006233819.1 | 3 - 4 | 539 | 90 | ||
XM_008761634.1 | XP_008759856.1 | 3 - 4 | 539 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |