Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769727.1 | XP_008767949.1 | 3 - 4 | 483 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |