Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751244.2 | XP_003751292.1 | 14 - 15 | 1171 | 78 | |
XM_003751245.2 | XP_003751293.1 | 15 - 16 | 1231 | 78 | ||
XM_006249933.2 | XP_006249995.1 | 14 - 15 | 1241 | 78 | ||
XM_006249934.1 | XP_006249996.1 | 15 - 16 | 1158 | 78 | ||
XM_008769580.1 | XP_008767802.1 | 13 - 14 | 1061 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |