Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100974.1 | NP_001094444.1 | 11 - 12 | 1785 | 56 | |
XM_006254807.2 | XP_006254869.1 | 11 - 12 | 1844 | 56 | ||
XM_006254808.2 | XP_006254870.1 | 11 - 12 | 3784 | 56 | ||
XM_006254809.2 | XP_006254871.1 | 11 - 12 | 1751 | 56 | ||
XM_006254811.2 | XP_006254873.1 | 11 - 12 | 1880 | 56 | ||
GenBank mRNA | AB199794.1 | - | 11 - 12 | 1785 | 56 | |
BC168236.1 | - | 1827 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |