Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047906.2 | NP_001041371.2 | 5 - 6 | 643 | 72 | |
GenBank mRNA | BC099199.1 | - | 5 - 6 | 617 | 7383928 | 72 |
FQ225106.1 | - | 5 - 6 | 623 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |