Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100520.1 | NP_001093990.1 | 4 - 5 | 1047 | 54 | |
GenBank mRNA | AY177754.1 | - | 4 - 5 | 807 | 54 | |
BC160852.1 | - | 4 - 5 | 1047 | 8364542 | 54 | |
FQ223245.1 | - | 742 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |