Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011949.1 | NP_001011949.1 | 5 - 6 | 1033 | 59 | |
XM_006232359.2 | XP_006232421.1 | 5 - 6 | 995 | 59 | ||
XM_006232360.2 | XP_006232422.1 | 5 - 6 | 998 | 59 | ||
XM_006232361.2 | XP_006232423.1 | 5 - 6 | 1038 | 59 | ||
XM_006232362.2 | XP_006232424.1 | 5 - 6 | 999 | 59 | ||
XM_008760987.1 | XP_008759209.1 | 5 - 6 | 1088 | 59 | ||
XM_008760988.1 | XP_008759210.1 | 5 - 6 | 979 | 59 | ||
GenBank mRNA | BC079234.1 | - | 5 - 6 | 1033 | 7132591 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |